siRNA Details
| Gene Symbol: 3110062M04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135611.1 | NP_001129083.1 | 2 | 332 | ||
| NM_199145.2 | NP_954596.1 | 2 | 657 | |||
| GenBank mRNA | AK042380.1 | - | 2 | 613 | ||
| AK083365.1 | - | 2 | 299 | |||
| AK170119.1 | - | 2 | 328 | |||
| BB611813.1 | - | 2 | 333 | |||
| BC029235.1 | - | 2 | 276 | 4912211 | ||

3110062M04Rik
Genomic Map