siRNA Details
| Gene Symbol: Pramel3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031390.2 | NP_113567.1 | 4 | 622 | ||
| GenBank mRNA | AY004873.1 | - | 4 | 636 | ||
| Gene Symbol: AV320801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177918.1 | NP_808586.1 | 623 | |||
| GenBank mRNA | AK029939.1 | - | 624 | |||
| Gene Symbol: Gm15023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099321.2 | NP_001092791.1 | 4 | 605 | ||
| GenBank mRNA | BC115763.1 | - | 4 | 605 | 40058793 | |
| BC119584.1 | - | 4 | 605 | 40058853 | ||
| BC119585.1 | - | 4 | 605 | 40058855 | ||

Pramel3
Genomic Map