siRNA Details
| Gene Symbol: Rhox2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114153.1 | NP_001107625.1 | 2 | 457 | ||
| Gene Symbol: Rhox2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029203.2 | NP_083479.1 | 2 | 441 | ||
| GenBank mRNA | AK015997.1 | - | 2 | 442 | ||
| BC132216.1 | - | 2 | 435 | 40130493 | ||
| BC132220.1 | - | 2 | 435 | 40130497 | ||
| BC145534.1 | - | 2 | 435 | |||
| DQ058639.1 | - | 2 | 442 | |||
| Gene Symbol: Rhox2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099316.1 | NP_001092786.1 | 2 | 419 | ||
| GenBank mRNA | BC049546.1 | - | 419 | |||
| Gene Symbol: Rhox2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099318.1 | NP_001092788.1 | 441 | |||
| Gene Symbol: Rhox2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081669.2 | NP_001075138.2 | 2 | 452 | ||
| Gene Symbol: Rhox2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085348.1 | NP_001078817.1 | 441 | |||

Rhox2g
Genomic Map