siRNA Details
| Gene Symbol: Krtap21-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028621.3 | NP_082897.2 | 1 | 673 | ||
| GenBank mRNA | AK003736.2 | - | 1 | 704 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap21-1| Gene Symbol: Krtap21-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028621.3 | NP_082897.2 | 1 | 673 | ||
| GenBank mRNA | AK003736.2 | - | 1 | 704 | ||