siRNA Details
| Gene Symbol: Ifi202b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008327.2 | NP_032353.2 | 5, 6 | 1359 | ||
| NM_011940.2 | NP_036070.2 | 5, 6 | 1356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifi202b| Gene Symbol: Ifi202b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008327.2 | NP_032353.2 | 5, 6 | 1359 | ||
| NM_011940.2 | NP_036070.2 | 5, 6 | 1356 | |||