siRNA Details
| Gene Symbol: Ifi202b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008327.2 | NP_032353.2 | 5 | 1270 | ||
| NM_011940.2 | NP_036070.2 | 5 | 1267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifi202b| Gene Symbol: Ifi202b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008327.2 | NP_032353.2 | 5 | 1270 | ||
| NM_011940.2 | NP_036070.2 | 5 | 1267 | |||