siRNA Details
| Gene Symbol: COBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287436.1 | NP_001274365.1 | 11 | 2510 | ||
| NM_015198.3 | NP_056013.2 | 10 | 2339 | |||
| XM_005271750.1 | XP_005271807.1 | 12 | 2546 | |||
| XM_005271751.1 | XP_005271808.1 | 11 | 2501 | |||
| XM_005271756.1 | XP_005271813.1 | 10 | 2330 | |||
| XM_011515234.1 | XP_011513536.1 | 13 | 2621 | |||
| XM_011515235.1 | XP_011513537.1 | 12 | 2576 | |||
| XM_011515236.1 | XP_011513538.1 | 13 | 2621 | |||
| XM_011515237.1 | XP_011513539.1 | 12 | 2450 | |||
| XM_011515238.1 | XP_011513540.1 | 13 | 2621 | |||
| XM_011515239.1 | XP_011513541.1 | 12 | 2450 | |||
| XM_011515240.1 | XP_011513542.1 | 11 | 2405 | |||
| XM_011515241.1 | XP_011513543.1 | 7 | 1625 | |||
| XM_017011898.1 | XP_016867387.1 | 11 | 2375 | |||
| XM_017011899.1 | XP_016867388.1 | 11 | 2501 | |||
| XM_017011900.1 | XP_016867389.1 | 10 | 2330 | |||
| GenBank mRNA | AB014533.1 | - | 10 | 2320 | ||
| AL713786.1 | - | 2 | 5794 | |||
| BC029275.1 | - | 5 | 1210 | |||
| BC045771.1 | - | 9 | 1809 | |||
| BC136441.1 | - | 11 | 2425 | 9020430 | ||
| BC144099.1 | - | 10 | 2254 | 9052619 | ||
| BC150263.1 | - | 10 | 2320 | 9007207 | ||
| CR936689.1 | - | 9 | 1830 | |||

COBL
Genomic Map