siRNA Details
| Gene Symbol: Grin3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033351.2 | NP_001028523.1 | 2 | 1404 | ||
| NM_001276355.1 | NP_001263284.1 | 2 | 1404 | |||
| GenBank mRNA | AK044171.1 | - | 2 | 1381 | ||
| AK138366.1 | - | 2 | 1405 | |||
| AK173314.1 | - | 2 | 405 | |||

Grin3a
Genomic Map