siRNA Details
| Gene Symbol: Tmem69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177670.4 | NP_808338.2 | 3 | 511 | ||
| GenBank mRNA | AK041960.1 | - | 3 | 496 | ||
| BC106178.1 | - | 3 | 504 | 6512571 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem69| Gene Symbol: Tmem69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177670.4 | NP_808338.2 | 3 | 511 | ||
| GenBank mRNA | AK041960.1 | - | 3 | 496 | ||
| BC106178.1 | - | 3 | 504 | 6512571 | ||