siRNA Details
| Gene Symbol: Otog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013624.2 | NP_038652.2 | 38 | 6340 | ||
| GenBank mRNA | U96411.1 | - | 39 | 6337 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otog| Gene Symbol: Otog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013624.2 | NP_038652.2 | 38 | 6340 | ||
| GenBank mRNA | U96411.1 | - | 39 | 6337 | ||