siRNA Details
| Gene Symbol: Greb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083628.1 | NP_001077097.1 | 3, 4 | 381 | ||
| GenBank mRNA | AK142556.1 | - | 3, 4 | 384 | ||
| BC147570.1 | - | 2, 3 | 238 | 9056560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Greb1l| Gene Symbol: Greb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083628.1 | NP_001077097.1 | 3, 4 | 381 | ||
| GenBank mRNA | AK142556.1 | - | 3, 4 | 384 | ||
| BC147570.1 | - | 2, 3 | 238 | 9056560 | ||