siRNA Details
| Gene Symbol: Vmn2r49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105156.1 | NP_001098626.1 | 3 | 1175 | ||
| Gene Symbol: Vmn2r32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105063.1 | NP_001098533.1 | 3 | 1175 | ||
| GenBank mRNA | AF011415.1 | - | 788 | |||
More Information
Set Membership: |
|

Vmn2r49
Genomic Map