siRNA Details
| Gene Symbol: ASTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184734.1 | NP_001171663.1 | 7 | 1117 | ||
| NM_014010.4 | NP_054729.3 | 21 | 3673 | |||
| NM_198186.3 | NP_937829.3 | 7 | 1438 | |||
| NM_198187.3 | NP_937830.3 | 7 | 1117 | |||
| NM_198188.2 | NP_937831.1 | 7 | 1117 | |||
| GenBank mRNA | AB014534.1 | - | 21 | 3673 | ||
| AF116574.1 | - | 19 | 2894 | |||
| AK024064.1 | - | 7 | 1088 | |||
| AK127350.1 | - | 20 | 2894 | |||
| AK315025.1 | - | 19 | 2737 | |||
| BC018759.2 | - | 15 | 2122 | |||
| BC029272.1 | - | 7 | 1438 | |||
| BC093835.1 | - | 7 | 951 | |||
| BC101667.1 | - | 7 | 950 | |||
| BC143289.1 | - | 7 | 951 | |||
| BC143292.1 | - | 7 | 951 | |||
| BC143293.1 | - | 7 | 951 | |||
| BC146756.1 | - | 21 | 3673 | 9007192 | ||
| BX649176.1 | - | 21 | 3008 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_207109, NM_019514, AK041160, AF116573.

ASTN2
Genomic Map