siRNA Details
| Gene Symbol: Gm3696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC055874.1 | - | 1301 | 3491523 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm3696| Gene Symbol: Gm3696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC055874.1 | - | 1301 | 3491523 | ||