siRNA Details
| Gene Symbol: Hist1h3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145073.2 | NP_659539.1 | 1 | 441 | ||
| GenBank mRNA | BC125355.1 | - | 1 | 437 | 40129870 | |
| BC125357.1 | - | 1 | 437 | 40129872 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h3g| Gene Symbol: Hist1h3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145073.2 | NP_659539.1 | 1 | 441 | ||
| GenBank mRNA | BC125355.1 | - | 1 | 437 | 40129870 | |
| BC125357.1 | - | 1 | 437 | 40129872 | ||