siRNA Details
| Gene Symbol: Raet1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020030.2 | NP_064414.1 | 588 | |||
| GenBank mRNA | AF257520.1 | - | 4 | 590 | ||
| BC106190.1 | - | 588 | 30142928 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Raet1d| Gene Symbol: Raet1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020030.2 | NP_064414.1 | 588 | |||
| GenBank mRNA | AF257520.1 | - | 4 | 590 | ||
| BC106190.1 | - | 588 | 30142928 | |||