siRNA Details
| Gene Symbol: Olfr746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146298.2 | NP_666410.2 | 1 | 596 | ||
| Gene Symbol: Olfr748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011837.2 | NP_001011837.2 | 1 | 596 | ||
More Information
Set Membership: |
|

Olfr746
Genomic Map