siRNA Details
| Gene Symbol: Rps26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013765.2 | NP_038793.2 | 3 | 249 | ||
| GenBank mRNA | AK008301.1 | - | 3 | 245 | ||
| AK010689.2 | - | 3 | 244 | |||
| AK012722.1 | - | 3 | 247 | |||
| AK018702.1 | - | 3 | 245 | |||
| AV585706.1 | - | 2 | 203 | |||
| BC036987.1 | - | 2 | 235 | 4987659 | ||
| BC081452.1 | - | 2 | 222 | 6411109 | ||
| BC100456.1 | - | 3 | 236 | 6541446 | ||
| BU530648.1 | - | 3 | 250 | |||
| U67770.1 | - | 2 | 226 | |||

Rps26
Genomic Map