siRNA Details
| Gene Symbol: 9830147E19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242388.1 | NP_001229317.1 | 4 | 1058 | ||
| GenBank mRNA | AK141688.1 | - | 4 | 1073 | ||
| AK162659.1 | - | 3 | 914 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9830147E19Rik| Gene Symbol: 9830147E19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242388.1 | NP_001229317.1 | 4 | 1058 | ||
| GenBank mRNA | AK141688.1 | - | 4 | 1073 | ||
| AK162659.1 | - | 3 | 914 | |||