siRNA Details
| Gene Symbol: Rps29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009093.2 | NP_033119.1 | 2 | 268 | ||
| GenBank mRNA | AA217935.1 | - | 2 | 152 | ||
| AK002939.1 | - | 2 | 172 | |||
| AK012285.1 | - | 2 | 171 | |||
| AK224925.1 | - | 2 | 161 | |||
| BC024393.1 | - | 188 | 4986748 | |||
| BC051203.1 | - | 2 | 155 | 1476903 | ||
| BC055750.1 | - | 2 | 219 | |||
| L31609.1 | - | 2 | 152 | |||

Rps29
Genomic Map