siRNA Details
| Gene Symbol: Rps29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009093.2 | NP_033119.1 | 3 | 341 | ||
| GenBank mRNA | AA217935.1 | - | 3 | 225 | ||
| AK002939.1 | - | 3 | 245 | |||
| AK012285.1 | - | 3 | 244 | |||
| AK224925.1 | - | 3 | 234 | |||
| BC024393.1 | - | 261 | 4986748 | |||
| BC051203.1 | - | 3 | 228 | 1476903 | ||
| BC055750.1 | - | 3 | 292 | |||
| L31609.1 | - | 3 | 225 | |||

Rps29
Genomic Map