siRNA Details
| Gene Symbol: Hist1h3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145073.2 | NP_659539.1 | 1 | 264 | ||
| GenBank mRNA | BC125355.1 | - | 1 | 260 | 40129870 | |
| BC125357.1 | - | 1 | 260 | 40129872 | ||
| Gene Symbol: Hist1h3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013550.5 | NP_038578.2 | 1 | 269 | ||
| GenBank mRNA | AK006742.1 | - | 1 | 249 | ||
| AK018952.1 | - | 1 | 270 | |||
| BC115816.1 | - | 1 | 266 | 40105082 | ||
| Gene Symbol: Hist1h3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178203.2 | NP_835510.1 | 1 | 236 | ||
| Gene Symbol: Hist1h3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175653.2 | NP_783584.1 | 1 | 260 | ||
| GenBank mRNA | BC120800.1 | - | 1 | 341 | 40129723 | |
| BC120802.1 | - | 1 | 341 | 40129725 | ||
| Gene Symbol: Hist1h3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178204.2 | NP_835511.1 | 1 | 275 | ||
| GenBank mRNA | AK020421.1 | - | 276 | |||
| Gene Symbol: Hist1h3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178205.2 | NP_835512.1 | 1 | 236 | ||
| GenBank mRNA | BC127042.1 | - | 1 | 229 | ||
| Gene Symbol: Hist1h3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013548.4 | NP_038576.1 | 1 | 669 | ||
| GenBank mRNA | AK010121.1 | - | 1 | 258 | ||
| BC101954.1 | - | 1 | 669 | 40044770 | ||
| BC101955.1 | - | 1 | 669 | 40044769 | ||
| BC101956.1 | - | 1 | 669 | 40044765 | ||
| BC103549.1 | - | 1 | 669 | 40044771 | ||
| Gene Symbol: Hist1h3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178206.2 | NP_835513.1 | 1 | 267 | ||
| GenBank mRNA | BC107285.2 | - | 1 | 234 | ||
| Gene Symbol: Hist1h3i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178207.2 | NP_835514.1 | 1 | 263 | ||
| GenBank mRNA | AK155722.1 | - | 264 | |||
| BC107287.2 | - | 1 | 232 | |||
| BC116383.2 | - | 1 | 236 | 40101629 | ||
| BC127148.1 | - | 1 | 235 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008771686, XM_001057443, XM_006253979, XM_008771684.

Hist1h3g
Genomic Map