siRNA Details
Gene Symbol: Whsc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079952.1 | - | 3, 4 | 814 | ||
AK156746.1 | - | 6, 7 | 2053 | |||
AK170040.1 | - | 6, 7 | 2116 | |||
BC036336.1 | - | 5, 6 | 1645 | |||
BC064447.1 | - | 6, 7 | 2091 |
