siRNA Details
Gene Symbol: Whsc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156746.1 | - | 2 | 924 | ||
AK170040.1 | - | 2 | 987 | |||
AK172238.1 | - | 2 | 950 | |||
BC036336.1 | - | 1 | 516 | |||
BC064447.1 | - | 2 | 962 |

More Information
Other Species Targeted

