siRNA Details
| Gene Symbol: Pifo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200028.1 | NP_001186957.1 | 5 | 689 | ||
| NM_029604.3 | NP_083880.2 | 5 | 641 | |||
| GenBank mRNA | AK006407.1 | - | 5 | 642 | ||
| AW519725.1 | - | 4 | 353 | |||
| BC049757.1 | - | 5 | 690 | 6773491 | ||
| HM237137.1 | - | 5 | 531 | |||
| HM237138.1 | - | 5 | 689 | |||

Pifo
Genomic Map