siRNA Details
| Gene Symbol: Phip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081216.1 | NP_001074685.1 | 27 | 3392 | ||
| GenBank mRNA | AF310251.1 | - | 4 | 656 | ||
| AK162189.1 | - | 8 | 962 | |||
| BC037950.1 | - | 1 | 25 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phip| Gene Symbol: Phip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081216.1 | NP_001074685.1 | 27 | 3392 | ||
| GenBank mRNA | AF310251.1 | - | 4 | 656 | ||
| AK162189.1 | - | 8 | 962 | |||
| BC037950.1 | - | 1 | 25 | |||