siRNA Details
| Gene Symbol: OTUD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015207.1 | NP_056022.1 | 4 | 716 | ||
| XM_005245792.3 | XP_005245849.1 | 4 | 1399 | |||
| XM_005245793.3 | XP_005245850.1 | 4 | 1400 | |||
| XM_005245794.3 | XP_005245851.1 | 3 | 351 | |||
| GenBank mRNA | AB007928.3 | - | 4 | 693 | ||

OTUD3
Genomic Map