siRNA Details
| Gene Symbol: Slc2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031197.2 | NP_112474.2 | 7, 8 | 1072 | ||
| GenBank mRNA | AK005068.1 | - | 7, 8 | 1073 | ||
| AK165430.1 | - | 7, 8 | 1007 | |||
| BC034675.1 | - | 7, 8 | 991 | 4236331 | ||
| X15684.1 | - | 7, 8 | 1021 | |||
| X16986.1 | - | 7, 8 | 1011 | |||

Slc2a2
Genomic Map