siRNA Details
| Gene Symbol: 5031410I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207657.3 | NP_997540.2 | 5 | 1294 | ||
| GenBank mRNA | AK077239.1 | - | 1295 | |||
| Gene Symbol: Gm10471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177579.1 | NP_001171050.1 | 5 | 1312 | ||

5031410I06Rik
Genomic Map