siRNA Details
| Gene Symbol: 5031410I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207657.3 | NP_997540.2 | 5 | 1301 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5031410I06Rik| Gene Symbol: 5031410I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207657.3 | NP_997540.2 | 5 | 1301 | ||