siRNA Details
| Gene Symbol: Pdlim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114087.2 | NP_001107559.1 | 3 | 242 | ||
| NM_001114088.2 | NP_001107560.1 | 3 | 242 | |||
| NM_026131.4 | NP_080407.3 | 3 | 242 | |||
| GenBank mRNA | AK003019.1 | - | 3 | 199 | ||
| AK003338.1 | - | 3 | 233 | |||
| AK010141.1 | - | 3 | 231 | |||
| AK010339.1 | - | 3 | 233 | |||
| AK033091.1 | - | 234 | ||||
| AK049902.1 | - | 3 | 234 | |||
| AK077998.1 | - | 3 | 234 | |||
| AK128987.1 | - | 3 | 230 | |||
| AK142933.1 | - | 3 | 225 | |||
| AK167476.1 | - | 3 | 234 | |||
| BC049565.1 | - | 3 | 230 | 6588160 | ||
| BC052698.1 | - | 3 | 213 | |||
| BY056634.1 | - | 3 | 235 | |||

Pdlim7
Genomic Map