siRNA Details
| Gene Symbol: Trim59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025863.3 | NP_080139.3 | 3 | 747 | ||
| GenBank mRNA | AK012269.1 | - | 3 | 748 | ||
| AK016642.1 | - | 3 | 942 | |||
| BC006700.1 | - | 3 | 665 | 3501285 | ||
| BC025430.1 | - | 3 | 680 | 4017983 | ||
| BC055347.1 | - | 1 | 217 | |||
| CD541510.1 | - | 1 | 94 | |||

Trim59
Genomic Map