siRNA Details
| Gene Symbol: Trim59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025863.3 | NP_080139.3 | 3 | 848 | ||
| GenBank mRNA | AK016642.1 | - | 3 | 1043 | ||
| BC006700.1 | - | 3 | 766 | 3501285 | ||
| BC025430.1 | - | 3 | 781 | 4017983 | ||
| BC055347.1 | - | 1 | 318 | |||
| CD541510.1 | - | 1 | 195 | |||

Trim59
Genomic Map