siRNA Details
| Gene Symbol: Cep162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199316.2 | NP_955020.2 | 21, 22 | 3102 | ||
| GenBank mRNA | AK135482.1 | - | 21, 22 | 3101 | ||
| BC150998.1 | - | 21, 22 | 2907 | 9087911 | ||
| BC151006.1 | - | 21, 22 | 2907 | 9087919 | ||
| BC151104.1 | - | 21, 22 | 2907 | 9088017 | ||
| CK639077.1 | - | 3, 4 | 405 | |||
| CN456181.1 | - | 1, 2 | 115 | |||

Cep162
Genomic Map