siRNA Details
| Gene Symbol: Cep162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199316.2 | NP_955020.2 | 27 | 4408 | ||
| GenBank mRNA | BC028799.1 | - | 5 | 543 | ||
| BC150998.1 | - | 27 | 4213 | 9087911 | ||
| BC151006.1 | - | 27 | 4213 | 9087919 | ||
| BC151104.1 | - | 27 | 4213 | 9088017 | ||

Cep162
Genomic Map