siRNA Details
| Gene Symbol: Nol12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133800.3 | NP_598561.2 | 1 | 118 | ||
| GenBank mRNA | AK036891.1 | - | 1 | 61 | ||
| AK037035.1 | - | 1 | 112 | |||
| AK083539.1 | - | 120 | ||||
| AK167446.1 | - | 1 | 112 | |||
| AY940114.1 | - | 1 | 61 | |||
| BC013701.1 | - | 1 | 103 | 3483364 | ||
| BC094424.1 | - | 1 | 105 | 6819926 | ||

Nol12
Genomic Map