siRNA Details
| Gene Symbol: Spata17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028848.3 | NP_083124.1 | 2 | 289 | ||
| NM_029067.1 | NP_083343.1 | 2 | 289 | |||
| GenBank mRNA | AK050925.1 | - | 2 | 290 | ||
| AY747687.1 | - | 2 | 264 | |||
| BC089520.1 | - | 2 | 264 | 6774024 | ||

Spata17
Genomic Map