siRNA Details
| Gene Symbol: Gm14483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111037.1 | NP_001104507.1 | 1 | 487 | ||
| GenBank mRNA | BC087913.1 | - | 1 | 487 | 6705184 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14483| Gene Symbol: Gm14483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111037.1 | NP_001104507.1 | 1 | 487 | ||
| GenBank mRNA | BC087913.1 | - | 1 | 487 | 6705184 | |