siRNA Details
| Gene Symbol: Vmn2r32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105063.1 | NP_001098533.1 | 3 | 737 | ||
| GenBank mRNA | AF011415.1 | - | 350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r32| Gene Symbol: Vmn2r32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105063.1 | NP_001098533.1 | 3 | 737 | ||
| GenBank mRNA | AF011415.1 | - | 350 | |||