siRNA Details
| Gene Symbol: Vmn2r84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081448.1 | NP_001074917.1 | 3 | 736 | ||
| Gene Symbol: Vmn2r85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102602.1 | NP_001096072.1 | 3 | 736 | ||
| Gene Symbol: Vmn2r86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103365.1 | NP_001096835.1 | 3 | 736 | ||
| Gene Symbol: Vmn2r87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103366.1 | NP_001096836.1 | 3 | 736 | ||
More Information
Set Membership: |
|

Vmn2r84
Genomic Map