siRNA Details
| Gene Symbol: Spice1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144550.4 | NP_653133.3 | 15 | 2323 | ||
| GenBank mRNA | AK040970.1 | - | 1 | 269 | ||
| AK048789.1 | - | 15 | 2267 | |||
| AY099108.1 | - | 15 | 2603 | |||
| BC024956.1 | - | 7 | 1677 | |||
| BC052381.1 | - | 15 | 2277 | 5703283 | ||
| BC056960.1 | - | 15 | 2277 | 5703283 | ||

Spice1
Genomic Map