siRNA Details
| Gene Symbol: Xlr4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293676.1 | NP_001280605.1 | 9 | 946 | ||
| NM_001293677.1 | NP_001280606.1 | 9 | 899 | |||
| NM_001293678.1 | NP_001280607.1 | 9 | 923 | |||
| NM_001293679.1 | NP_001280608.1 | 9 | 935 | |||
| NM_001293680.1 | NP_001280609.1 | 9 | 959 | |||
| NM_021365.3 | NP_067340.2 | 9 | 1063 | |||
| GenBank mRNA | AL136330.1 | - | 9 | 1030 | ||
| BC025576.1 | - | 9 | 899 | 5323621 | ||
| BC034848.1 | - | 9 | 1487 | |||
| BC138975.1 | - | 9 | 805 | 8861997 | ||
| BC138979.1 | - | 9 | 829 | 8862001 | ||
| BC145481.1 | - | 9 | 829 | 9054044 | ||
| BC145482.1 | - | 9 | 841 | 9054045 | ||
| BC145483.1 | - | 9 | 865 | 9054046 | ||

Xlr4b
Genomic Map