siRNA Details
| Gene Symbol: Tbc1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081278.2 | NP_001074747.2 | 17 | 4011 | ||
| GenBank mRNA | AK047705.1 | - | 7 | 1182 | ||
| AK171689.1 | - | 6 | 1202 | |||
| AK172575.1 | - | 6 | 1082 | |||
| AK220347.1 | - | 17 | 2421 | |||

Tbc1d4
Genomic Map