siRNA Details
| Gene Symbol: Gm13154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014397.4 | NP_001014419.2 | 6 | 1815 | ||
| GenBank mRNA | BC085112.1 | - | 6 | 1797 | 30731985 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13154| Gene Symbol: Gm13154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014397.4 | NP_001014419.2 | 6 | 1815 | ||
| GenBank mRNA | BC085112.1 | - | 6 | 1797 | 30731985 | |