siRNA Details
| Gene Symbol: Oxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011025.4 | NP_035155.1 | 1, 2 | 157 | ||
| GenBank mRNA | AF483508.1 | - | 1, 2 | 116 | ||
| AF483509.1 | - | 1, 2 | 116 | |||
| AK003042.1 | - | 1, 2 | 154 | |||
| BC117030.1 | - | 1, 2 | 136 | 40126349 | ||
| BC117032.1 | - | 1, 2 | 136 | 40126351 | ||

Oxt
Genomic Map