siRNA Details
| Gene Symbol: AF366264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153093.3 | NP_694733.3 | 1 | 833 | ||
| GenBank mRNA | AK030169.1 | - | 1 | 399 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AF366264| Gene Symbol: AF366264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153093.3 | NP_694733.3 | 1 | 833 | ||
| GenBank mRNA | AK030169.1 | - | 1 | 399 | ||