siRNA Details
| Gene Symbol: Vps13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177184.3 | NP_796158.2 | 47 | 5597 | ||
| GenBank mRNA | AB281675.1 | - | 47 | 5597 | ||
| AK049557.1 | - | 9 | 1238 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps13c| Gene Symbol: Vps13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177184.3 | NP_796158.2 | 47 | 5597 | ||
| GenBank mRNA | AB281675.1 | - | 47 | 5597 | ||
| AK049557.1 | - | 9 | 1238 | |||