siRNA Details
Gene Symbol: Hist4h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175652.3 | NP_783583.1 | 1 | 299 | ||
GenBank mRNA | AK045187.1 | - | 294 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hist4h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175652.3 | NP_783583.1 | 1 | 299 | ||
GenBank mRNA | AK045187.1 | - | 294 |