siRNA Details
| Gene Symbol: Defa-rs10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007845.3 | NP_031871.1 | 148 | |||
| GenBank mRNA | BC119615.1 | - | 148 | 40061719 | ||
| BC119616.1 | - | 148 | 40061721 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defa-rs10| Gene Symbol: Defa-rs10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007845.3 | NP_031871.1 | 148 | |||
| GenBank mRNA | BC119615.1 | - | 148 | 40061719 | ||
| BC119616.1 | - | 148 | 40061721 | |||