siRNA Details
| Gene Symbol: Cyhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276322.1 | NP_001263251.1 | 3 | 1157 | ||
| NM_001276333.1 | NP_001263262.1 | 2 | 1200 | |||
| NM_001276334.1 | NP_001263263.1 | 3 | 984 | |||
| NM_001276335.1 | NP_001263264.1 | 3 | 930 | |||
| NM_180962.2 | NP_851293.1 | 3 | 1223 | |||
| GenBank mRNA | AK004009.1 | - | 3 | 1067 | ||
| BC016196.1 | - | 3 | 1021 | 4488866 | ||
| BC025112.1 | - | 3 | 1123 | 3670832 | ||

Cyhr1
Genomic Map